missing translation for 'onlineSavingsMsg'
Learn More

1-Isopropyl-1H-1,2,3-benzotriazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10622453
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10622453 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10622453 Supplier Thermo Scientific Maybridge Supplier No. KM10480DA
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 306935-41-1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H11N3O2
MDL Number MFCD01570664
Quantity 1 g
Synonym 1-isopropyl-1h-1,2,3-benzotriazole-5-carboxylic acid, 1-isopropyl-1h-benzo d 1,2,3 triazole-5-carboxylic acid, 1-isopropyl-1,2,3-benzotriazole-5-carboxylic acid, 1-isopropyl-1h-benzotriazole-5-carboxylic acid, 1h-benzotriazole-5-carboxylic acid, 1-1-methylethyl, 1-isopropylbenzotriazole-5-carboxylic acid, 1h-benzotriazole-5-carboxylicacid, 1-1-methylethyl, 1-propan-2-yl-1h-1,2,3-benzotriazole-5-carboxylic acid, 1-methylethyl benzotriazole-5-carboxylic acid, maybridge1_005244
InChI Key RUTVRAJKELSHCC-UHFFFAOYSA-N
SMILES CC(C)N1N=NC2=CC(=CC=C12)C(O)=O
IUPAC Name 1-(propan-2-yl)-1H-1,2,3-benzotriazole-5-carboxylic acid
Molecular Weight (g/mol) 205.22
PubChem CID 2736690
Formula Weight 205.22
Percent Purity 97%
Chemical Name or Material 1-Isopropyl-1H-1, 2, 3-benzotriazole-5-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.