missing translation for 'onlineSavingsMsg'
Learn More

1-Isopropylpiperazine, 98+%

Product Code. 10300584 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10300584 5 g Glass bottle 5g
10530954 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10300584 Supplier Thermo Scientific Acros Supplier No. 384810050
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4318-42-7 | C7H16N2 | 128.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4318-42-7
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.22
MDL Number MFCD00167971
InChI Key WHKWMTXTYKVFLK-UHFFFAOYSA-N
Synonym 1-isopropylpiperazine, n-isopropylpiperazine, 1-propan-2-yl piperazine, piperazine, 1-isopropyl, isopropyl piperazine, 1-isopropyl-piperazine, 1-1-methylethyl piperazine, n-isopropyl piperazine, 1-isopropyl piperazine, piperazine, 1-1-methylethyl
PubChem CID 78013
IUPAC Name 1-propan-2-ylpiperazine
SMILES CC(C)N1CCNCC1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Yellow
Density 0.8960g/mL
Boiling Point 180°C to 181°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4700 to 1.472
Quantity 5 g
Specific Gravity 0.896
Formula Weight 128.22
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 1-Isopropylpiperazine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Isopropylpiperazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.