missing translation for 'onlineSavingsMsg'
Learn More

1-Mercaptoadamantane, 98%

Product Code. 15465128
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15465128 250 mg 250mg
15455128 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465128 Supplier Thermo Scientific Alfa Aesar Supplier No. H58958.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34301-54-7 | C10H16S | 168.298 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34301-54-7
Molecular Formula C10H16S
Molecular Weight (g/mol) 168.298
MDL Number MFCD00213530
InChI Key ADJJLNODXLXTIH-UHFFFAOYSA-N
Synonym 1-adamantanethiol, adamantanethiol, 1-adamantyl hydrosulfide, tricyclo 3.3.1.13,7 decane-1-thiol, tricyclo 3.3.1.1 3,7 decane-1-thiol, acmc-20aoyp, 1-admsh, pubchem21526, 1-adsh
PubChem CID 99730
IUPAC Name adamantane-1-thiol
SMILES C1C2CC3CC1CC(C2)(C3)S

Specifications

Melting Point 99°C to 106°C
Quantity 250 mg
UN Number UN2811
Sensitivity Hygroscopic
Formula Weight 168.3
Percent Purity 98%
Chemical Name or Material 1-Mercaptoadamantane
compliance-icons compliance-icons
Nombre del producto
  • 1-Mercaptoadamantane
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Indicaciones de peligro
  • H301-Toxic if swallowed.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.