missing translation for 'onlineSavingsMsg'
Learn More

1-(Mercaptomethyl)cyclopropaneacetic acid, 98%, Thermo Scientific™

Product Code. 15448288
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448288 5 g 5g
15438288 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448288 Supplier Thermo Scientific Alfa Aesar Supplier No. H61723.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162515-68-6
Molecular Formula C6H10O2S
Molecular Weight (g/mol) 146.204
MDL Number MFCD03411600
InChI Key VFAXPOVKNPTBTM-UHFFFAOYSA-N
Synonym 2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropaneacetic acid, 1-mercaptomethyl cyclopropylacetic acid, 1-mercaptomethyl-cyclopropaneacetic acid, unii-1x00t93wyx, cyclopropaneacetic acid, 1-mercaptomethyl, 2-1-sulfanylmethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropyl acetic acid, 1-sulfanylmethyl cyclopropyl acetic acid
PubChem CID 9793825
IUPAC Name 2-[1-(sulfanylmethyl)cyclopropyl]acetic acid
SMILES C1CC1(CC(=O)O)CS

Specifications

Melting Point 42°C to 46°C
Color White
Flash Point >110°C (230°F)
Odor Stench
Quantity 5 g
UN Number UN3261
Formula Weight 146.21
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1-(Mercaptomethyl)cyclopropaneacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.