missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-2-propanol, 98.5%, extra pure

Product Code. 10296950 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
5 L
25 L
Packaging:
Glass Bottle
Metal drum
Plastic drum
Unit Size:
1 L
2.5 L
25 L
25mL
5 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10296950 2.5 L Glass Bottle 2.5 L
10531101 25 mL Glass Bottle 25mL
10057252 1 L Glass Bottle 1 L
10746433 5 L Plastic drum 5 L
11924741 25 L Metal drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10296950 Supplier Thermo Scientific Acros Supplier No. 244990025
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 107-98-2 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-98-2
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00004537
InChI Key ARXJGSRGQADJSQ-UHFFFAOYNA-N
Synonym 1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b
PubChem CID 7900
IUPAC Name 1-methoxypropan-2-ol
SMILES COCC(C)O

Specifications

Melting Point -97.0°C
Density 0.9200g/mL
Boiling Point 120.0°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass Bottle
Linear Formula CH3OCH2CH(OH)CH3
Refractive Index 1.4020 to 1.4040
Quantity 2.5 L
Beilstein 01,536
Residue after Evaporation 0.01% max.
Specific Gravity 0.92
Solubility Information Solubility in water: 1000g/L (20°C)
Viscosity 1.7 mPa.s (20°C)
Formula Weight 90.12
Percent Purity 98.5%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 1-Methoxy-2-propanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Methoxy-2-propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.