missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-2-propanol, 99+%

Product Code. 11353316
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
4 x 4 L
Unit Size:
1 L
16 L
250mL
4 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11353316 250 mL 250mL
11394298 250 mL 250mL
11304308 1 L 1 L
11363316 1 L 1 L
11314308 4 L 4 L
11324308 4 x 4 L 16 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11353316 Supplier Thermo Scientific Alfa Aesar Supplier No. L12694.AK
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 107-98-2 | C4H10O2 | 90.12 g/mol

1-Methoxy-2-propanol is used as a reagent in the synthesis of 2-amino-3-carboxy-4-phenylthiophenes, which acts as a protein kinase C inhibitors. It is also used as a reagent in the synthesis of metolachlor. It acts as a good biological indicator. Further, it is used as a solvent and an antifreeze agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 107-98-2
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00004537
InChI Key ARXJGSRGQADJSQ-UHFFFAOYNA-N
Synonym 1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b
PubChem CID 7900
IUPAC Name 1-methoxypropan-2-ol
SMILES COCC(C)O

Spezifikation

Melting Point -100°C
Color Colorless
Density 0.926 g/mL
pH 4 to 7
Boiling Point 119°C to 120°C
Flash Point 35°C
Assay Percent Range >99%
Odor Alcohol-like
Refractive Index 1.403
Quantity 250 mL
UN Number 3092
Beilstein 1731270
Solubility Information Miscible with water,methanol and ether.
Formula Weight 90.12
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material 1-Methoxy-2-propanol
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • 1-Methoxy-2-propanol
Signalwort
  • Danger
Gefahrenkategorie
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H226-Flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Ergänzende Informationen
  • MIXTURE LIST-Contain: 1-Methoxy-2-propanol

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.