missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-2-propanol, TRC

Product Code. 30134468
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134468 10 g 10g
30134231 50 g 50g
30005979 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134468 Supplier Toronto Research Chemicals Supplier No. TRCM26500510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-98-2
Molecular Formula C4 H10 O2
Molecular Weight (g/mol) 90.12
Synonym 2-Propanol, 1-methoxy- (6CI, 7CI, 8CI, 9CI, ACI), 1-Methoxy-2-propanol (ACI), 1-Methoxy-2-hydroxypropane, 2-Methoxy-1-methylethanol, 3-Methoxy-2-propanol, Closol, Dowanol 33B, Dowtherm 209, Essilor altius, Glycol ether PM, Methoxyisopropanol, NSC 2409, Propylene glycol 1-methyl ether
IUPAC Name 1-methoxypropan-2-ol
SMILES COCC(C)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H10O2/c1-4(5)3-6-2/h4-5H,3H2,1-2H3
Formula Weight 90.0681
Chemical Name or Material 1-Methoxy-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.