missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1,2,4-triazole, 98%

Product Code. 11374955
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374955 5 g 5g
11364955 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374955 Supplier Thermo Scientific Alfa Aesar Supplier No. L18337.06
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6086-21-1 | C3H5N3 | 83.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6086-21-1
Molecular Formula C3H5N3
Molecular Weight (g/mol) 83.09
MDL Number MFCD01076192
InChI Key MWZDIEIXRBWPLG-UHFFFAOYSA-N
Synonym 1-methyl-1h-1,2,4-triazole, 1h-1,2,4-triazole, 1-methyl, 1-methyl,-1,2,4-triazole, 1-methyl-1h-1,2,4 triazole, pubchem22534, acmc-1avow, ksc495k1j
PubChem CID 22459
SMILES CN1C=NC=N1

Specifications

Melting Point 16°C
Density 1.1
Boiling Point 177°C to 178°C
Flash Point 81°C (177°F)
Refractive Index 1.4705
Quantity 5 g
Beilstein 106245
IUPAC Name 1-methyl-1H-1,2,4-triazole
Formula Weight 83.1
Percent Purity 99%
Chemical Name or Material 1-Methyl-1,2,4-triazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.