missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1-cyclohexene, 97%, stabilized

Product Code. 10665813 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10665813 10 g 10g
10444643 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10665813 Supplier Thermo Scientific Acros Supplier No. 156840100
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 591-49-1 | C7H12 | 96.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-49-1
InChI Key CTMHWPIWNRWQEG-UHFFFAOYSA-N
Synonym 1-methyl-1-cyclohexene, cyclohexene, 1-methyl, 1-methylcyclohex-1-ene, cyclohexene, methyl, methylcyclohexene, .alpha.-methylcyclohexene, 1-methyl-cyclohexene, methyl-1-cyclohexene, 2,3,4,5-tetrahydrotoluene, unii-te4p8q2044
PubChem CID 11574
IUPAC Name 1-methylcyclohexene
SMILES CC1=CCCCC1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless
Density 0.8100g/mL
Boiling Point 110.0°C to 111.0°C
Flash Point −3°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Molecular Formula C7H12
Linear Formula C6H9CH3
Refractive Index 1.4492 to 1.4512
Quantity 10 g
Beilstein 05, 66
Specific Gravity 0.81
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,acetone and benzene
Stabilizer 10ppm hydroquinone
Molecular Weight (g/mol) 96.17
Formula Weight 96.17
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 1-Methyl-1-cyclohexene, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Methyl-1-cyclohexene, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.