missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-imidazol-2-yl)methanol, 97%, Thermo Scientific™

Product Code. 10672433
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10672433 250 mg Amber Glass Bottle 250mg
10562662 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10672433 Supplier Thermo Scientific Maybridge Supplier No. CC20809CB
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 17334-08-6
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.132
MDL Number MFCD00964673
InChI Key CDQDMLWGTVLQEE-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazol-2-yl methanol, 1-methylimidazol-2-yl methanol, 1h-imidazole-2-methanol, 1-methyl, 1-methyl-2-imidazolyl methanol, 2-hydroxymethyl-1-methylimidazole, imidazole-2-methanol, 1-methyl, 1-methyl-1h-imidazole-2-methanol, 1-methylimidazol-2-yl methan-1-ol, pubchem9047
PubChem CID 573612
IUPAC Name (1-methylimidazol-2-yl)methanol
SMILES CN1C=CN=C1CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material (1-Methyl-1H-imidazol-2-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.