missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-imidazol-5-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10576524
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10576524 5 g Amber Glass Bottle 5g
10197562 250 mg Amber Glass Bottle 250mg
10573143 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10576524 Supplier Thermo Scientific Maybridge Supplier No. CC23613DE
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 486414-86-2
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.148
InChI Key PYAQTQXFMQWCHQ-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazol-5-yl methylamine, 1-methyl-5-aminomethylimidazole, 1-methyl-1h-imidazol-5-yl methanamine, 3-methylimidazol-4-yl methanamine, 1h-imidazole-5-methanamine, 1-methyl, 5-aminomethyl-1-methyl-1h-imidazole, c-3-methyl-3h-imidazol-4-yl-methylamine, 1-3-methylimidazol-4-yl methanamine, 1-methyl-imidazol-5-yl methylamine, 1-1-methyl-1h-imidazole-5-yl methanamine
PubChem CID 2795424
IUPAC Name (3-methylimidazol-4-yl)methanamine
SMILES CN1C=NC=C1CN

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 111.15
Percent Purity 97%
Chemical Name or Material (1-Methyl-1H-imidazol-5-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.