missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-4-carbonitrile, 97%, Thermo Scientific™

Product Code. 10675135
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
Product Code. Quantity Packaging unitSize
10675135 5 g Amber Glass Bottle 5g
10610315 250 mg Amber Glass Bottle 250mg
10620315 1 g Amber Glass Bottle 1g
3 options
This item is not returnable. View return policy

Product Code. 10675135

Brand: Thermo Scientific Maybridge CC03616DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 66121-69-5
Molecular Formula C5H5N3
Molecular Weight (g/mol) 107.116
MDL Number MFCD06411537
InChI Key LAKVILSENFFPRR-UHFFFAOYSA-N
PubChem CID 2795115
IUPAC Name 1-methylimidazole-4-carbonitrile
SMILES CN1C=C(N=C1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 1-methyl-1h-imidazole-4-carbonitrile, 4-cyano-1-methyl-1h-imidazole, 1h-imidazole-4-carbonitrile, 1-methyl, 4-cyano-1-methylimidazole, 1-methyl-4-cyano-1h-imidazole, 1-methyl-4-imidazolecarbonitrile, 1h-imidazole-4-carbonitrile,1-methyl
Formula Weight 107.12
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-imidazole-4-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.