missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-4-sulfonamide, 97%, Thermo Scientific™

Product Code. 10213822
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10213822 250 mg Amber Glass Bottle 250mg
10724053 1 g Amber Glass Bottle 1g
10215294 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10213822 Supplier Thermo Scientific Maybridge Supplier No. TL01003CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 111124-90-4
Molecular Formula C4H7N3O2S
Molecular Weight (g/mol) 161.18
MDL Number MFCD02091382
InChI Key QBJSSOBNVYDCDQ-UHFFFAOYSA-N
PubChem CID 2736885
IUPAC Name 1-methylimidazole-4-sulfonamide
SMILES CN1C=NC(=C1)S(N)(=O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 1-methyl-1h-imidazole-4-sulfonamide, 1h-imidazole-4-sulfonamide,1-methyl, 1-methyl-1h-imidazole-4-sulphonamide, acmc-20dtvv, maybridge1_000050, spr_7, 1-methyl-4-imidazolesulfonamide, 1-methyl-1h-imidazol-4-ylsulfonamide, 1h-imidazole-4-sulfonamide, 1-methyl
Formula Weight 161.18
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-imidazole-4-sulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.