missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10502482
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10502482 1 g Amber Glass Bottle 1g
10566524 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10502482 Supplier Thermo Scientific Maybridge Supplier No. CC23604DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 39021-62-0
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
InChI Key BNYKZFOZWZMEJD-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-5-carbaldehyde, 1-methyl-1h-imidazole-5-carboxaldehyde, 3-methyl-3h-imidazole-4-carbaldehyde, 1-methylimidazole-5-carboxaldehyde, 1h-imidazole-5-carboxaldehyde, 1-methyl, 1-methylimidazole-5-carbaldehyde, 1-methyl-5-imidazolecarboxaldehyde, 1-methylimidazole-5-carboxaldeyde, pubchem8047, 5-formyl-1-methylimidazole
PubChem CID 573592
IUPAC Name 3-methylimidazole-4-carbaldehyde
SMILES CN1C=NC=C1C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 110.12
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-imidazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.