missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10702493
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10702493 1 g 1g
10200355 5 g 5g
10779215 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10702493 Supplier Thermo Scientific Maybridge Supplier No. CC23601DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 41806-40-0
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD00955677
InChI Key PBEDVTDUVXFSMW-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-5-carboxylic acid, 1-methylimidazole-5-carboxylic acid, 5-carboxy-1-methyl-1h-imidazole, 3-methyl-3h-imidazole-4-carboxylic acid, 1-methyl-1h-imidazole-5-carboxylicacid, 1-methyl-1h-5-imidazolecarboxylic acid, pubchem20954, acmc-209jlu, ksc236i7p, 1-methyl-5-imidazolecarboxylic acid
PubChem CID 2773507
IUPAC Name 3-methylimidazole-4-carboxylic acid
SMILES CN1C=NC=C1C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-imidazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.