missing translation for 'onlineSavingsMsg'
Learn More

1-methyl-1h-indazole-3-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10215014
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215014 10 g Amber Glass Bottle 10g
10015822 250 mg Amber Glass Bottle 250mg
10105512 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10215014 Supplier Thermo Scientific Maybridge Supplier No. CC06302EA
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 106649-02-9
Molecular Formula C9H7ClN2O
Molecular Weight (g/mol) 194.62
MDL Number MFCD02093096
InChI Key NPNWVMBPSINFBV-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazole-3-carbonyl chloride, 1h-indazole-3-carbonylchloride, 1-methyl, 1-methyl-1h-indazol-3-carbonyl chloride, 1h-indazole-3-carbonyl chloride, 1-methyl, acmc-20mac8, 1-methyl-3-indazolylcarbonyl chloride, 1-methyl-indazole-3-carbonyl chloride, 1-methyl-1h-indazole-3-carboxychloride, 1-methyl-1h-indazole-3-carboxylic acid chloride
PubChem CID 7022055
IUPAC Name 1-methylindazole-3-carbonyl chloride
SMILES CN1N=C(C(Cl)=O)C2=CC=CC=C12

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 194.62
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indazole-3-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.