missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-indol-5-yl)methanol, 97%, Thermo Scientific™

Product Code. 10691236
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10691236 5 g Amber Glass Bottle 5g
10399853 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10691236 Supplier Thermo Scientific Maybridge Supplier No. CC41409DE
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 448967-90-6
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.20
MDL Number MFCD09025838
InChI Key CEBLTQGXYITWTM-UHFFFAOYSA-N
Synonym 1-methyl-1h-indol-5-yl methanol, 1-methylindol-5-yl methanol, 1h-indole-5-methanol, 1-methyl-9ci
PubChem CID 24229490
IUPAC Name (1-methylindol-5-yl)methanol
SMILES CN1C=CC2=CC(CO)=CC=C12

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 161.2
Percent Purity 97%
Chemical Name or Material (1-Methyl-1H-indol-5-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.