missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-4-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10412834
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10412834 10 g Amber Glass Bottle 10g
10327594 250 mg Amber Glass Bottle 250mg
10103674 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10412834 Supplier Thermo Scientific Supplier No. CC45803EA
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 876316-36-8
Molecular Formula C9H8ClNO2S
Molecular Weight (g/mol) 229.678
MDL Number MFCD08690260
InChI Key HFWFOJFQXZBLAU-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole-4-sulfonyl chloride, 1h-indole-4-sulfonylchloride, 1-methyl, 1-methyl-1h-indole-4-sulphonyl chloride, chloro 1-methylindol-4-yl sulfone, 1-methyl-1h-indole-4-sulfonylchloride, 4-chlorosulphonyl-1-methyl-1h-indole, 1h-indole-4-sulfonyl chloride, 1-methyl
PubChem CID 18525782
IUPAC Name 1-methylindole-4-sulfonyl chloride
SMILES CN1C=CC2=C1C=CC=C2S(=O)(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 229.69
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indole-4-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.