missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10442284
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10442284 250 mg 250mg
10418043 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10442284 Supplier Thermo Scientific Maybridge Supplier No. CC41404CB
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 90923-75-4
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD08271908
InChI Key XIVDZBIBWGQOTI-UHFFFAOYSA-N
PubChem CID 7537534
SMILES CN1C=CC2=CC(C=O)=CC=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 1-methyl-1h-indole-5-carbaldehyde, 1h-indole-5-carboxaldehyde, 1-methyl, 5-formyl-1-methylindole, n-methylindol-5-carbaldehyde, n-methylindole-5-carbaldehyde, n-methylindole-5-carboxaldehyde, 1-methyl-1h-indole-5-carboxaldehyde, 1h-indole-5-carboxaldehyde,1-methyl
IUPAC Name 1-methyl-1H-indole-5-carbaldehyde
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.