missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-6-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10718075
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10718075 1 g Amber Glass Bottle 1g
10173374 250 mg Amber Glass Bottle 250mg
10392195 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10718075 Supplier Thermo Scientific Maybridge Supplier No. CC44204DA
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 21005-45-8
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD08690251
InChI Key QGAKKGIWURKKHV-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole-6-carbaldehyde, 1h-indole-6-carboxaldehyde, 1-methyl, 1h-indole-6-carboxaldehyde,1-methyl, 6-formy-1-methylindole, 6-formyl-1-methylindole, 1-methyl-1h-indole-6-carboxaldehyde
PubChem CID 14835679
IUPAC Name 1-methylindole-6-carbaldehyde
SMILES CN1C=CC2=CC=C(C=O)C=C12

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indole-6-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.