missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-7-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10032034
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10032034 250 mg Amber Glass Bottle 250mg
10397304 1 g Amber Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10032034

Brand: Thermo Scientific Maybridge CC66904CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 69047-36-5
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD11109329
InChI Key JOBIAZYEJOMXEP-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole-7-carbaldehyde, 1h-indole-7-carboxaldehyde, 1-methyl, 1-methyl-7-formylindole, n-methylindole-7-carboxaldehyde, jobiazyejomxep-uhfffaoysa, 1-methyl-1h-indole-7-carboxaldehyde, 1h-indole-7-carboxaldehyde, 1-methyl-9ci
PubChem CID 11040979
IUPAC Name 1-methylindole-7-carbaldehyde
SMILES CN1C=CC2=C1C(=CC=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indole-7-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.