missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazol-5-ylamine, 97%, Thermo Scientific™

Product Code. 10533192
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10533192 1 g 1g
10543192 10 g 10g
10103464 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10533192 Supplier Thermo Scientific Maybridge Supplier No. GK03066DA
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1192-21-8
Molecular Formula C4H7N3
Molecular Weight (g/mol) 97.121
MDL Number MFCD00068156
InChI Key JESRNIJXVIFVOV-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazol-5-amine, 1-methyl-1h-pyrazol-5-ylamine, 1-methyl-5-aminopyrazole, 5-amino-1-methylpyrazole, 5-amino-1-methyl-1h-pyrazole, 1h-pyrazol-5-amine, 1-methyl, 2-methyl-2h-pyrazol-3-ylamine, 5-amino-1-methyl-pyrazole, 2-methyl-2h-pyrazol-3-amine, 1-methylpyrazole-5-ylamine
PubChem CID 136927
IUPAC Name 2-methylpyrazol-3-amine
SMILES CN1C(=CC=N1)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 97.12
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrazol-5-ylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.