missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazol-5-ylamine, 97%, Thermo Scientific™

Product Code. 10543192
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10543192 10 g 10g
10533192 1 g 1g
10103464 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10543192 Supplier Thermo Scientific Maybridge Supplier No. GK03066EA
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kemiska identifierare

CAS 1192-21-8
Molecular Formula C4H7N3
Molecular Weight (g/mol) 97.121
MDL Number MFCD00068156
InChI Key JESRNIJXVIFVOV-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazol-5-amine, 1-methyl-1h-pyrazol-5-ylamine, 1-methyl-5-aminopyrazole, 5-amino-1-methylpyrazole, 5-amino-1-methyl-1h-pyrazole, 1h-pyrazol-5-amine, 1-methyl, 2-methyl-2h-pyrazol-3-ylamine, 5-amino-1-methyl-pyrazole, 2-methyl-2h-pyrazol-3-amine, 1-methylpyrazole-5-ylamine
PubChem CID 136927
IUPAC Name 2-methylpyrazol-3-amine
SMILES CN1C(=CC=N1)N

Specifikationer

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 97.12
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrazol-5-ylamine
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.