missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-3-carboxylic acid, 96%

Product Code. 11449480
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449480 1 g 1g
11439480 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449480 Supplier Thermo Scientific Alfa Aesar Supplier No. H50317.03
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 25016-20-0 | C5H6N2O2 | 126.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25016-20-0
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00464254
InChI Key YBFIKNNFQIBIQZ-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl, 3-carboxy-1-methyl-1h-pyrazole, zlchem 1152, acmc-209ggi, 1-methylpyrazolecarboxylic acid, 1-methyl-pyrazole-3-carboxylic acid, pyrazole-3-carboxylic acid, 1-methyl, 1h-pyrazole-3-carboxylicacid, 1-methyl, 1-methyl-1h-4-pyrazole-3-carboxylic acid
PubChem CID 573176
ChEBI CHEBI:74738
IUPAC Name 1-methylpyrazole-3-carboxylic acid
SMILES CN1C=CC(=N1)C(O)=O

Specifications

Melting Point 146°C to 156°C
Odor Odorless
Quantity 1 g
Formula Weight 126.11
Percent Purity 96%
Chemical Name or Material 1-Methyl-1H-pyrazole-3-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.