missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-4-boronic acid hydrochloride, 95%, Thermo Scientific™

Product Code. 11986241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11986241 1 g 1g
11976241 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11986241 Supplier Thermo Scientific Acros Supplier No. 445490010
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1026796-02-0
Molecular Formula C4H8BClN2O2
Molecular Weight (g/mol) 162.38
MDL Number MFCD09972098
InChI Key YFPLZINTZVVVJM-UHFFFAOYSA-N
Synonym 1-methylpyrazole-4-boronic acid, hcl, 1-methyl-1h-pyrazol-4-yl boronic acid hydrochloride, 1-methyl-1h-pyrazole-4-boronic acid hydrochloride, 1-methylpyrazole-4-boronic acid hcl, 1-methyl-1h-pyrazol-4-ylboronic acid hydrochloride, 1-methylpyrazol-4-ylboronic acid hydrochloride, 1-methylpyrazole-4-boronic acid,hcl, 1-methyl-1h-pyrazole-4-boronic acid xhcl, 1-methylpyrazole-4-boronic acid hydrochloride, 1-methyl-1h-pyrazole-4-boronic acid hcl
PubChem CID 45158882
IUPAC Name (1-methylpyrazol-4-yl)boronic acid;hydrochloride
SMILES Cl.CN1C=C(C=N1)B(O)O

Specifications

Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 162.38
Percent Purity 95%
Chemical Name or Material 1-Methyl-1H-pyrazole-4-boronic acid hydrochloride
compliance-icons
Product Identifier
  • 1-Methyl-1H-pyrazole-4-boronic acid hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.