missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 15450416
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450416 1 g 1g
15460416 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450416 Supplier Thermo Scientific Alfa Aesar Supplier No. H53139.03
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 761446-44-0 | Molecular Formula: C10H17BN2O2 | Molecular Weight: 208.07 g/mol

It is a reagent used for Suzuki-Miyaura cross-coupling reactions, transesterification reactions. It is involved in several reactions as a reagent for the preparation of aminothiazoles as γ-secretase modulators,amino-pyrido-indol-carboxamides, as potential JAK2 inhibitors for myeloproliferative disorders therapy, pyridine derivatives as TGF-β1 and active A signaling inhibitors and MK-2461 analogs as inhibitors of c-Met kinase for the treatment of cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reagent used for Suzuki-Miyaura cross-coupling reactions, transesterification reactions. It is involved in several reactions as a reagent for the preparation of aminothiazoles as γ-secretase modulators,amino-pyrido-indol-carboxamides, as potential JAK2 inhibitors for myeloproliferative disorders therapy, pyridine derivatives as TGF-β1 and active A signaling inhibitors and MK-2461 analogs as inhibitors of c-Met kinase for the treatment of cancer.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 761446-44-0
Molecular Formula C10H17BN2O2
Molecular Weight (g/mol) 208.07
MDL Number MFCD03789259
InChI Key UCNGGGYMLHAMJG-UHFFFAOYSA-N
Synonym 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-methylpyrazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-methyl-1h-pyrazole-4-boronic acid pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-methyl-pyrazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-methyl-4-pyrazoleboronic acid pinacol ester
PubChem CID 2773987
IUPAC Name 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 59°C to 64°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 208.07
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrazole-4-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.