missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-4-carboxaldehyde, 96%

Product Code. 15450518
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450518 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15450518 Supplier Thermo Scientific Alfa Aesar Supplier No. H54146.03
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 25016-11-9 | C5H6N2O | 110.116 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 25016-11-9
Melting Point 29°C to 34°C
Flash Point >230°C (446°F)
Molecular Formula C5H6N2O
MDL Number MFCD00460465
Quantity 1 g
Sensitivity Air Sensitive
Synonym 1-methyl-1h-pyrazole-4-carbaldehyde, 1-methyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 1-methyl, 4-formyl-1-methylpyrazole, 4-formyl-1-methyl-1h-pyrazole, 1-methylpyrazole-4-carboxaldehyde, 1-methylpyrazole-4-carboxyaldehyde, 1-methyl-1h-pyrazol-4-carbaldehyd, 1-methylpyrazole-4-aldehyde, acmc-1cs06
InChI Key MYFZXSOYJVWTBL-UHFFFAOYSA-N
SMILES CN1C=C(C=N1)C=O
IUPAC Name 1-methylpyrazole-4-carbaldehyde
Molecular Weight (g/mol) 110.116
PubChem CID 573117
Formula Weight 110.12
Percent Purity 96%
Chemical Name or Material 1-Methyl-1H-pyrazole-4-carboxaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.