missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-5-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10702125
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10702125 1 g Amber Glass Bottle 1g
10327264 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10702125 Supplier Thermo Scientific Maybridge Supplier No. CC62303DA
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1020721-61-2
Molecular Formula C4H5ClN2O2S
Molecular Weight (g/mol) 180.606
MDL Number MFCD09065021
InChI Key OWLKLUGPVSESBX-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-5-sulfonyl chloride, 2-methyl-2h-pyrazole-3-sulfonyl chloride, 5-chlorosulphonyl-1-methyl-1h-pyrazole, chloro 1-methylpyrazol-5-yl sulfone, 1-methyl-1h-pyrazole-5-sulphonyl chloride, 2-methylpyrazol-3-yl methanesulfonyl chloride
PubChem CID 22472621
IUPAC Name 2-methylpyrazole-3-sulfonyl chloride
SMILES CN1C(=CC=N1)S(=O)(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 180.61
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrazole-5-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.