missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole, 97+%

Product Code. p-7046537
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11303706

Brand: Thermo Scientific Alfa Aesar L14831.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 930-36-9 | C4H6N2 | 82.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-36-9
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.106
MDL Number MFCD00144943
InChI Key UQFQONCQIQEYPJ-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole, n-methylpyrazole, 1h-pyrazole, 1-methyl, pyrazole, 1-methyl, methylpyrazole, 1-methyl pyrazole, 1-methypyrazole, n-methyl pyrazole, 1-methyl-pyrazole, zlchem 255
PubChem CID 70255
ChEBI CHEBI:59025
IUPAC Name 1-methylpyrazole
SMILES CN1C=CC=N1

Specifications

Boiling Point 127°C
Flash Point 35°C (95°F)
Odor Pungent
Quantity 1 g
UN Number UN1993
Beilstein 105207
Formula Weight 82.11
Percent Purity ≥97%
Chemical Name or Material 1-Methyl-1H-pyrazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.