missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrrole-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10220115
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10220115 5 g 5g
10255074 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10220115 Supplier Thermo Scientific Maybridge Supplier No. CC02701DE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 6973-60-0
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD00003088
InChI Key ILAOVOOZLVGAJF-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrrole-2-carboxylic acid, 1-methyl-2-pyrrolecarboxylic acid, n-methylpyrrole-2-carboxylic acid, 1h-pyrrole-2-carboxylic acid, 1-methyl, 1h-pyrrolecarboxylic acid, methyl, 1-methyl-1h-pyrrole-2-carboxylicacid, pubchem9181, acmc-209oar, ksc353i0t, 2-carboxy-1-methyl-1h-pyrrole
PubChem CID 81453
IUPAC Name 1-methylpyrrole-2-carboxylic acid
SMILES CN1C=CC=C1C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 125.13
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrrole-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.