missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-2-imidazolemethanol, 98%
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Description
1-Methyl-2-imidazolemethanol is used as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Methyl-2-imidazolemethanol is used as intermediates.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid, acid chlorides, halogens, halocarbons.
1-Methyl-2-imidazolemethanol is used as intermediates.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid, acid chlorides, halogens, halocarbons.
Specifications
Specifications
| Melting Point | 107°C to 111°C |
| Quantity | 25 g |
| Beilstein | 113492 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 112.13 |
| Percent Purity | 98% |
| Physical Form | Solid |
| Chemical Name or Material | 1-Methyl-2-imidazolemethanol |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction