missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-imidazolidinone, 98+%

Product Code. 10523231 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2 g
Packaging:
Glass bottle
Unit Size:
2g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10523231 500 mg Glass bottle 500mg
10554664 2 g Glass bottle 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10523231 Supplier Thermo Scientific Acros Supplier No. 304155000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 694-32-6 | C4H8N2O | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 694-32-6
Molecular Formula C4H8N2O
Molecular Weight (g/mol) 100.12
MDL Number MFCD00800601
InChI Key JTPZTKBRUCILQD-UHFFFAOYSA-N
PubChem CID 567600
IUPAC Name 1-methylimidazolidin-2-one
SMILES CN1CCNC1=O

Specifica

CAS Min % 98.0
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 500 mg
Synonym 1-methyl-2-imidazolidinone, 2-imidazolidinone, 1-methyl, 1-methyl-2-imidazolidone, n-methylimidazolidinone, acmc-209yb9, 3-methylimidazolidin-2-one, 1-methylimidazolidine-2-one, 1-methyl-imidazolidin-2-one, 2-imidazolidinone,1-methyl, 1-methyl-2-imidazolidinone #
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 100.12
Percent Purity 98+%
Physical Form Crystalline Solid
Chemical Name or Material 1-Methyl-2-imidazolidinone, 98+%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.