missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-phenylindole, 99%

Product Code. 11449617
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449617 250 g 250g
11429617 10 g 10g
11439617 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449617 Supplier Thermo Scientific Alfa Aesar Supplier No. B22105.30
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3558-24-5 | C15H13N | 207.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 3558-24-5
Molecular Formula C15H13N
Molecular Weight (g/mol) 207.276
MDL Number MFCD00022892
InChI Key SFWZZSXCWQTORH-UHFFFAOYSA-N
Synonym 1-methyl-2-phenyl-1h-indole, 2-phenyl-n-methylindole, 1h-indole, 1-methyl-2-phenyl, n-methyl-2-phenylindole, indole, 1-methyl-2-phenyl, pubchem7435, n-methyl-2-phenyl indole, n-methyl-2-phenyl-indole, acmc-1cp6x, 1-methyl-2-phenyl-indole
PubChem CID 77095
IUPAC Name 1-methyl-2-phenylindole
SMILES CN1C2=CC=CC=C2C=C1C3=CC=CC=C3

Spécification

Melting Point 98°C to 99°C
Quantity 250 g
Beilstein 148770
Formula Weight 207.28
Percent Purity 99%
Chemical Name or Material 1-Methyl-2-phenylindole

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.