Product Code. 10466622
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Glass Bottle
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
10466622
1 L
AcroSeal™ Glass Bottle
1 L
10229441
100 mL
AcroSeal™ Glass Bottle
100mL
10533864
2.5 L
Glass Bottle
2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code.
10466622
Supplier
Thermo Scientific Acros
Supplier No.
326930010
180.00 € 174.00 € / 1 L Save 6.00 € 3% Off
Estimated Shipment: 06-10-2026Log in to see stock.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
CAS: 872-50-4 | C5H9NO | 99.133 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
CAS
872-50-4
Molecular Formula
C5H9NO
Molecular Weight (g/mol)
99.133
MDL Number
MFCD00003193
InChI Key
SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID
13387
ChEBI
CHEBI:7307
IUPAC Name
1-methylpyrrolidin-2-one
SMILES
CN1CCCC1=O
Specifications
Melting Point
-24.0°C
CAS Min %
99.4
Color
Colorless
CAS Max %
100.0
Density
1.0300g/mL
Quantity
1 L
Flash Point
91°C
Infrared Spectrum
Authentic
Packaging
AcroSeal™ Glass Bottle
Physical Form
Liquid
Refractive Index
1.4690 to 1.4710
Assay Percent Range
99.5%
Specific Gravity
1.03
Viscosity
1.67 mPa.s (20°C)
Water
0.005% Max. (Coulometric)
Beilstein
21, 237
Fieser
01,696; 02,281; 09,316; 11,346
Merck Index
15, 6190
Boiling Point
202.0°C
Solubility Information
Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight
99.13
Percent Purity
99.5%
Grade
Extra Dry
Chemical Name or Material
1-Methyl-2-pyrrolidinone
Show More
Show Less
Product Identifier
Signal Word
Hazard Category
Skin corrosion/irritation Category 2
Serious eye damage/eye irritation Category 2
Reproductive toxicity Category 1B
Specific target organ toxicity Category 3
Hazard Statement
H315-Causes skin irritation.
H319-Causes serious eye irritation.
H335-May cause respiratory irritation.
H360D-May damage the unborn child.
Precautionary Statement
P302+P352-IF ON SKIN: Wash with plenty of water/soap.
P337+P313-If eye irritation persists: Get medical advice/attention.
P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
P312-Call a POISON CENTER/doctor/if you feel unwell.
P280-Wear protective gloves/protective clothing/eye protection/face protection.
P332+P313-If skin irritation occurs: Get medical advice/attention.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?