missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-Pyrrolidinone, 99%, Reagent Grade, Solstice

Product Code. 15647970 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
18 L
25 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
1 L
18 L
25 L
Pack of 4
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15647970 6 x 1 L Glass Bottle Pack of 6
15607970 1 L Glass Bottle 1 L
15637970 4 x 4 L Glass Bottle Pack of 4
15697960 18 L Stainless Steel Can 18 L
15627970 25 L Stainless Steel Can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15647970 Supplier Fluka Supplier No. M796036X1L
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD00003193
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

pH 8.5 to 10 (at 100.00g/L 20°C)
Melting Point -24°C
Color Colorless
Density 1.028g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 0.095
Flash Point 91°C (195.8°F)
Ignition Point 518°F
Odor Amine-like
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 0.99mmHg (40°C), 0.29mmHg (20°C)
Beilstein 106420
Boiling Point 204°C (1013 hPa)
Solubility Information Fully miscible in water
Formula Weight 99.13g/mol
Percent Purity 0.99
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Methyl-2-pyrrolidinone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: N-Methyl-2-pyrrolidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.