missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone-d9, 97% (Isotopic), Thermo Scientific™

Product Code. 11394418
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394418 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11394418 Supplier Thermo Scientific Alfa Aesar Supplier No. 042336.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

NMR Solvent

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Flash Point 86°C (187°F)
Quantity 1 g
CAS 185964-60-7
Melting Point -24°C
Boiling Point 81°C to 82°C (10mmHg)
Molecular Formula C5H9NO
MDL Number MFCD00148961
Synonym n-methyl-d3-2-pyrrolidinone-d6, 1-∼2∼h_3_ methyl ∼2∼h_6_ pyrrolidin-2-one, 1-methyl-2-pyrrolidinone-d9, 1-methyl-d3-2-pyrrolidinone-d6, 1-methyl-2-pyrrolidinone-d isotopic, n-methyl-d3-2-pyrrolidinone-d6, 98 atom % d, 1-2 h? methyl 2 h? pyrrolidin-2-one
Solubility Information Soluble in water,chloroform(unlabeled),and methanol(unlabeled).
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
SMILES CN1CCCC1=O
IUPAC Name 3,3,4,4,5,5-hexadeuterio-1-(trideuteriomethyl)pyrrolidin-2-one
Molecular Weight (g/mol) 99.13
PubChem CID 16217555
Formula Weight 108.21
Percent Purity 97%
Chemical Name or Material 1-Methyl-2-pyrrolidinone-d9, Isotopic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Informativa sulla privacy.