missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, Reagent Grade, Solstice

Product Code. 15617970
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15617970 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15617970 Supplier Fluka Supplier No. M796032.5L
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

CAS 872-50-4
Density 1.028g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.095
Flash Point 91°C (195.8°F)
Ignition Point 518°F
Molecular Formula C5H9NO
Vapor Pressure 0.99 mmHg (40°C), 0.29 mmHg (20°C)
MDL Number MFCD00003193
Beilstein 106420
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
SMILES CN1CCCC1=O
IUPAC Name 1-methylpyrrolidin-2-one
Molecular Weight (g/mol) 99.133
PubChem CID 13387
ChEBI CHEBI:7307
Formula Weight 99.13g/mol
Percent Purity 99%
Grade Reagent
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.