missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-(tri-n-butylstannyl)pyrrole, 98%, Thermo Scientific Chemicals

Product Code. 15478087
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15478087 5 g 5g
15488087 250 mg 250mg
15468087 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15478087 Supplier Thermo Scientific Alfa Aesar Supplier No. H51438.06
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 118486-97-8 | Molecular Formula: C17H33NSn | Molecular Weight: 370.168 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118486-97-8
Molecular Formula C17H33NSn
Molecular Weight (g/mol) 370.168
MDL Number MFCD01319022
InChI Key DINAKCGOEKXDTP-UHFFFAOYSA-N
Synonym 1-methyl-2-tributylstannyl pyrrole, 1-methyl-2-tributylstannyl-1h-pyrrole, 1-methyl-2-tributylstannylpyrrole, 1h-pyrrole,1-methyl-2-tributylstannyl, 1-methylpyrrol-2-yl tributyltin, 1-methyl-2-tri-n-butylstannyl pyrrole, n-methyl-2-tributylstannyl-1h-pyrrole, acmc-20aotm, tributyl-1-methylpyrrol-2-yl stannane, tributylstannyl-1-methylpyrrole
PubChem CID 2769397
IUPAC Name tributyl-(1-methylpyrrol-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CN1C

Specifications

Flash Point 98°C (208°F)
Quantity 5 g
UN Number UN2788
Formula Weight 370.15
Percent Purity 98%
Chemical Name or Material 1-Methyl-2-(tri-n-butylstannyl)pyrrole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.