missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-n-octylimidazolium chloride, 97%

Product Code. 15455888
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455888 50 g 50g
15445888 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455888 Supplier Thermo Scientific Alfa Aesar Supplier No. H59534.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 64697-40-1 | C12H23ClN2 | 230.78 g/mol

Role of 1-methyl-3-octylimidazolium chloride in the micellization behavior of amphiphilic drug amitriptyline hydrochloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Role of 1-methyl-3-octylimidazolium chloride in the micellization behavior of amphiphilic drug amitriptyline hydrochloride.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64697-40-1
Molecular Formula C12H23ClN2
Molecular Weight (g/mol) 230.78
MDL Number MFCD03095432
InChI Key OXFBEEDAZHXDHB-UHFFFAOYSA-M
Synonym 1-methyl-3-n-octylimidazolium chloride, 1-methyl-3-octylimidazolium chloride, 1-octyl-3-methylimidazolium chloride, 3-methyl-1-octylimidazolium chloride, omimcl, acmc-20aj1j, dsstox_cid_27849, dsstox_rid_82602, dsstox_gsid_47873, ksc491c7j
PubChem CID 2734223
ChEBI CHEBI:61342
IUPAC Name 1-methyl-3-octylimidazol-1-ium;chloride
SMILES [Cl-].CCCCCCCCN1C=C[N+](C)=C1

Specifications

Color Yellow
Density 1.01 g/mL
Refractive Index 1.51
Quantity 50 g
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 230.78
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-n-octylimidazolium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.