missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-octylimidazolium chloride, 97%, ACROS Organics™

Product Code. 11926261 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11926261 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11926261 Supplier Thermo Scientific Acros Supplier No. 356610500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Ionic liquid
TRUSTED_SUSTAINABILITY

Specifications

CAS 64697-40-1
Assay Percent Range 97%
Molecular Formula C12H23ClN2
MDL Number MFCD03095432
Quantity 50 g
Synonym 1-methyl-3-n-octylimidazolium chloride, 1-methyl-3-octylimidazolium chloride, 1-octyl-3-methylimidazolium chloride, 3-methyl-1-octylimidazolium chloride, omimcl, acmc-20aj1j, dsstox_cid_27849, dsstox_rid_82602, dsstox_gsid_47873, ksc491c7j
InChI Key OXFBEEDAZHXDHB-UHFFFAOYSA-M
SMILES [Cl-].CCCCCCCCN1C=C[N+](C)=C1
IUPAC Name 3-methyl-1-octyl-1H-imidazol-3-ium chloride
Molecular Weight (g/mol) 230.78
PubChem CID 2734223
ChEBI CHEBI:61342
Formula Weight 230.78
Chemical Name or Material 1-Methyl-3-octylimidazolium chloride
Show More Show Less
compliance-icons
Product Identifier
  • 1-Methyl-3-octylimidazolium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.