missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-phenyl-1H-pyrazol-5-amine, 97%, Thermo Scientific™

Product Code. 10242882
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10242882 1 g 1g
10672443 10 g 10g
10673455 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10242882 Supplier Thermo Scientific Maybridge Supplier No. KM01136DA
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10199-50-5
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.219
MDL Number MFCD00067874
InChI Key KCYRMURRLLYLPU-UHFFFAOYSA-N
Synonym 1-methyl-3-phenyl-1h-pyrazol-5-amine, 5-amino-1-methyl-3-phenylpyrazole, 5-amino-1-methyl-3-phenyl-1h-pyrazole, 2-methyl-5-phenyl-2h-pyrazol-3-ylamine, 1-methyl-3-phenylpyrazole-5-ylamine, acmc-1bxci, maybridge1_004465, 1-methyl-3-phenyl-5-pyrazolamine, 1-methyl-3-phenyl-pyrazol-5-amine, 2-methyl-5-phenyl-pyrazol-3-amine
PubChem CID 517779
IUPAC Name 2-methyl-5-phenylpyrazol-3-amine
SMILES CN1C(=CC(=N1)C2=CC=CC=C2)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-phenyl-1H-pyrazol-5-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.