missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-3-phenyl-1H-pyrazol-5-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10562864
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10562864 250 mg 250mg
10376284 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10562864 Supplier Thermo Scientific Maybridge Supplier No. CC46613CB
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 876728-39-1
Molecular Formula C11H13N3
Molecular Weight (g/mol) 187.25
MDL Number MFCD08271943
InChI Key XAVNJYHSYZUYFI-UHFFFAOYSA-N
Synonym 1-methyl-3-phenyl-1h-pyrazol-5-yl methylamine, 1-methyl-3-phenyl-1h-pyrazol-5-yl methanamine, 2-methyl-5-phenylpyrazol-3-yl methanamine, 1h-pyrazole-5-methanamine,1-methyl-3-phenyl, 1-1-methyl-3-phenyl-1h-pyrazol-5-yl methanamine, 1-methyl-3-phenylpyrazol-5-yl methylamine, 1-2-methyl-5-phenylpyrazol-3-yl methanamine, 5-aminomethyl-1-methyl-3-phenyl-1h-pyrazole, 5-aminomethyl-1-methyl-1h-pyrazol-3-yl benzene
PubChem CID 20109911
SMILES CN1N=C(C=C1CN)C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
IUPAC Name 1-(1-methyl-3-phenyl-1H-pyrazol-5-yl)methanamine
Formula Weight 187.24
Percent Purity 97%
Chemical Name or Material (1-Methyl-3-phenyl-1H-pyrazol-5-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.