missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-phenyl-1H-pyrazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10583874
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10583874 250 mg Amber Glass Bottle 250mg
10051054 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10583874 Supplier Thermo Scientific Maybridge Supplier No. CC46601CB
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10250-64-3
Molecular Formula C11H10N2O2
Molecular Weight (g/mol) 202.213
MDL Number MFCD08271939
InChI Key RTBQZMPYPFQBHA-UHFFFAOYSA-N
PubChem CID 7537625
IUPAC Name 2-methyl-5-phenylpyrazole-3-carboxylic acid
SMILES CN1C(=CC(=N1)C2=CC=CC=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 1-methyl-3-phenyl-1h-pyrazole-5-carboxylic acid, 2-methyl-5-phenyl-2h-pyrazole-3-carboxylic acid, 1-methyl-3-phenylpyrazole-5-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1-methyl-3-phenyl, 5-carboxy-1-methyl-3-phenyl-1h-pyrazole, 1-methyl-3-phenyl-1h-5-pyrazolecarboxylic acid, 1h-pyrazole-5-carboxylicacid, 1-methyl-3-phenyl, 1-methyl-3-phenyl-1 h-pyrazole-5-carboxylic acid
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-phenyl-1H-pyrazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.