missing translation for 'onlineSavingsMsg'
Learn More

1-methyl-3-trifluoromethylpyrazole-5-boronic acid, 97%, Thermo Scientific™

Product Code. 10347594
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10347594 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10347594 Supplier Thermo Scientific Maybridge Supplier No. MAY00027DA
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 344591-91-9
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C5H6BF3N2O2
MDL Number MFCD08701785
Quantity 1 g
Synonym 1-methyl-3-trifluoromethylpyrazole-5-boronic acid, 1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl boronic acid, 1-methyl-3-trifluoromethyl-1h-pyrazole-5-boronic acid, 5-borono-1-methyl-3-trifluoromethyl-1h-pyrazole, 2-methyl-5-trifluoromethyl pyrazol-3-ylboronic acid, 3-trifluoromethyl-1-methyl-1h-pyrazol-5-yl-5-boronic acid, 1-methyl-3-trifluoromethyl-1h-pyrazol-5-ylboronic acid, boronic acid, 1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl, acmc-209i7r
InChI Key XPUNXPPXMANQGF-UHFFFAOYSA-N
SMILES B(C1=CC(=NN1C)C(F)(F)F)(O)O
IUPAC Name [2-methyl-5-(trifluoromethyl)pyrazol-3-yl]boronic acid
Molecular Weight (g/mol) 193.92
PubChem CID 12056713
Formula Weight 193.92
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-trifluoromethylpyrazole-5-boronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.