missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-(4-piperidinyl)piperazine, 98%

Product Code. 11357219
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357219 1 g 1g
11367219 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357219 Supplier Thermo Scientific Alfa Aesar Supplier No. H27408.03
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 53617-36-0 | C10H21N3 | 183.30 g/mol

It is used both as a reagent and building block in several synthetic applications. As intermediate. As an excellent catalyst for many condensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used both as a reagent and building block in several synthetic applications. As intermediate. As an excellent catalyst for many condensation reactions.

Solubility
Soluble in water.

Notes
Air Sensitive & Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water, moisture, acids. Store under dry inert gas. Refrigerated (2-4°C).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53617-36-0
Molecular Formula C10H21N3
Molecular Weight (g/mol) 183.30
MDL Number MFCD03274729
InChI Key MRYYJGQKVGZGSB-UHFFFAOYSA-N
Synonym 1-methyl-4-piperidin-4-yl piperazine, 1-methyl-4-4-piperidinyl piperazine, 1-methyl-4-piperidin-4-yl-piperazine, piperazine, 1-methyl-4-4-piperidinyl, 4-methyl-1-4-piperidyl piperazine, acmc-1b13b, 1-methyl-4-4-piperidyl piperazine, 1-methyl-4-piperidin-4yl-piperazine, 1-methyl-4-4-piperidino piperazine
PubChem CID 795707
SMILES CN1CCN(CC1)C1CCNCC1

Specifications

Melting Point 53°C to 56°C
Quantity 1 g
UN Number UN3259
Sensitivity Air sensitive; Hygroscopic
Solubility Information Soluble in water.
IUPAC Name 1-methyl-4-(piperidin-4-yl)piperazine
Formula Weight 183.3
Percent Purity 98%
Chemical Name or Material 1-Methyl-4-(4-piperidinyl)piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.