missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-piperidone, 98%

Product Code. 11450983
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450983 50 g 50g
11460983 100 g 100g
11474160 250 g 250g
11464160 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11450983 Supplier Thermo Scientific Alfa Aesar Supplier No. A13352.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1445-73-4 | C6H11NO | 113.16 g/mol

1-Methyl-4-piperidone, are widely used in artificial fibre industry. They are polymerizable and used as nylon precursors. They have excellent applications as solvents and as intermediates for organic synthesis. They are used in the aqueous carrier medium and amide penetrants in inks and water soluble paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methyl-4-piperidone, are widely used in artificial fibre industry. They are polymerizable and used as nylon precursors. They have excellent applications as solvents and as intermediates for organic synthesis. They are used in the aqueous carrier medium and amide penetrants in inks and water soluble paints.

Solubility
It is miscible with water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents, strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1445-73-4
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00006191
InChI Key HUUPVABNAQUEJW-UHFFFAOYSA-N
Synonym 1-methyl-4-piperidone, n-methyl-4-piperidone, 1-methyl-4-piperidinone, 4-piperidinone, 1-methyl, 4-piperidone, 1-methyl, n-methyl-4-piperidinone, 1-methyl-4-piperidione, n-methylpiperidine-4-one, 1-methyl-4-oxopiperidine, 1-methylpiperidine-4-one
PubChem CID 74049
IUPAC Name 1-methylpiperidin-4-one
SMILES CN1CCC(=O)CC1

Specifications

Density 0.973
pH 12
Boiling Point 55°C to 58°C (10 mmHg)
Flash Point 60°C (140°F)
Odor Amine-like
Refractive Index 1.461
Quantity 50 g
UN Number UN2733
Beilstein 106924
Sensitivity Air sensitive
Solubility Information It is miscible with water
Formula Weight 113.16
Percent Purity 98%
Chemical Name or Material 1-Methyl-4-piperidone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Methyl-4-piperidone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1-Methyl-4-piperidone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.