missing translation for 'onlineSavingsMsg'
Learn More

[1-Methyl-5-bis(2’-hydroxyethyl)aminobenzimidazolyl-2]butanoic Acid Ethyl Ester, TRC

Product Code. 30478157
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478157 1 g 1g
30473006 100 mg 100mg
30465173 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478157 Supplier Toronto Research Chemicals Supplier No. TRCM2941901G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3543-74-6
Molecular Formula C18 H27 N3 O4
Molecular Weight (g/mol) 349.42
Synonym 5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Ethyl Ester, [1-Methyl-5-bis(2'-hydroxyethyl)aminobenzimidazolyl-2]butanoic Acid Ethyl Ester, 4-[5-[Bis(2-hydroxyethyl)amino]-1-methylbenzimidazol-2-yl]butanoic acid ethyl ester, Ethyl 5-[bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoate
IUPAC Name ethyl 4-[5-[bis(2-hydroxyethyl)amino]-1-methylbenzimidazol-2-yl]butanoate
SMILES CCOC(=O)CCCc1nc2cc(ccc2n1C)N(CCO)CCO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H27N3O4/c1-3-25-18(24)6-4-5-17-19-15-13-14(7-8-16(15)20(17)2)21(9-11-22)10-12-23/h7-8,13,22-23H,3-6,9-12H2,1-2H3
Formula Weight 349.2002
Chemical Name or Material 5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.