missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-5-thien-2-yl-1H-pyrazol-3-yl)methanol, 97%, Thermo Scientific™

Product Code. 11370647
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11370647 250 mg 250mg
10366374 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11370647 Supplier Thermo Scientific Maybridge Supplier No. CC47109CB
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 879896-47-6
Molecular Formula C9H10N2OS
Molecular Weight (g/mol) 194.252
MDL Number MFCD08435882
InChI Key HJYKWWXWCFOUID-UHFFFAOYSA-N
Synonym 1-methyl-5-thien-2-yl-1h-pyrazol-3-yl methanol, 1h-pyrazole-3-methanol,1-methyl-5-2-thienyl, 1-methyl-5-thiophen-2-yl pyrazol-3-yl methanol, 1-methyl-5-thiophen-2-yl-1h-pyrazol-3-yl methanol, 1-methyl-5-2-thienyl pyrazol-3-yl methan-1-ol, 3-hydroxymethyl-1-methyl-5-2-thienyl-1h-pyrazole
PubChem CID 18525806
IUPAC Name (1-methyl-5-thiophen-2-ylpyrazol-3-yl)methanol
SMILES CN1C(=CC(=N1)CO)C2=CC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 194.26
Percent Purity 97%
Chemical Name or Material (1-Methyl-5-thien-2-yl-1H-pyrazol-3-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.