missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10310953
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10310953 250 mg 250mg
10065762 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10310953 Supplier Thermo Scientific Maybridge Supplier No. GK00761CB
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 113100-53-1
Molecular Formula C6H5F3N2O2
Molecular Weight (g/mol) 194.113
MDL Number MFCD01936005
InChI Key FZNKJQNEJGXCJH-UHFFFAOYSA-N
Synonym 1-methyl-3-trifluoromethyl-1h-pyrazole-4-carboxylic acid, 1-methyl-3-trifluoromethyl pyrazole-4-carboxylic acid, 3-trifluoromethyl-1-methyl-1h-pyrazole-4-carboxylic acid, 1-methyl-3-trifluoromethyl-4-pyrazolecarboxylic acid, 1-methyl-3-trifluoromethyl-1h-pyrazole-4-carboxy, 1h-pyrazole-4-carboxylic acid, 1-methyl-3-trifluoromethyl, pubchem11402, acmc-1bscj, ksc183q3j
PubChem CID 2775665
IUPAC Name 1-methyl-3-(trifluoromethyl)pyrazole-4-carboxylic acid
SMILES CN1C=C(C(=N1)C(F)(F)F)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 194.11
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.