missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-nicotinamide Methosulphate, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Methyl-nicotinamide Methosulphate |
| CAS | 58971-09-8 |
| Molecular Formula | C7 H9 N2 O . C H3 O4 S |
| Quantity | 2 g |
| Synonym | Pyridinium','3-(aminocarbonyl)-1-methyl-','methyl sulfate (1:1), Pyridinium','3-(aminocarbonyl)-1-methyl-','methyl sulfate (9CI), 1-Methyl-nicotinamide methosulphate, 1-Methylnicotinamide methosulfate, N-Methylnicotinamide methosulfate |
| InChI Formula | InChI=1S/C7H8N2O.CH4O4S/c1-9-4-2-3-6(5-9)7(8)10;1-5-6(2,3)4/h2-5H,1H3,(H-,8,10);1H3,(H,2,3,4) |
| SMILES | COS(=O)(=O)[O-].C[n+]1cccc(c1)C(=O)N |
| IUPAC Name | 1-methylpyridin-1-ium-3-carboxamide;methyl sulfate |
| Molecular Weight (g/mol) | 248.26 |
| Formula Weight | 248.0467 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?